VeriSpec™ Chloride - Cl⁻ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010911-100N
RV010911-250N
RV010911-500N
RCRV010911100NEA
173.8
CAD
RCRV010911100N
RCRV010911250N
RCRV010911500N
VeriSpec™ Chloride - Cl⁻ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards
Anionic Standards
Chloride Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility