VeriSpec® Cesium - Cs⁺ Standard for Ion Chromatography 1000 ppm in H₂O
Supplier: Ricca Chemical
RV010908-100N RV010908-250N RV010908-500N
RCRV010908100NEA 170.64 CAD
RCRV010908100N RCRV010908250N RCRV010908500N
VeriSpec® Cesium - Cs⁺ Standard for Ion Chromatography 1000 ppm in H₂O
Standards Cationic Standards Cesium Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility