VeriSpec® Multi-Element Standard, Components, RICCA Chemical Company
Supplier: Ricca Chemical
RV010763-100N RV010763-500N
RCRV010763100NEA 922.72 CAD
RCRV010763100N RCRV010763500N
VeriSpec® Multi-Element Standard, Components, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
Certified reference material (CRM) for ICP-MS
50 ppm: As, K; 10 ppm: La, Li, Mn, Ni, Sr, Zn; 1 ppm Ba, Mg
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility