VeriSpec® Mixed Cation Standard, 6 components, Ricca Chemical Company
Supplier: Ricca Chemical
RV010689-50N RV010689-100N
RCRV010689-50NEA 425.02 CAD
RCRV010689-50N RCRV010689100N
VeriSpec® Mixed Cation Standard, 6 components, Ricca Chemical Company
Standards Cationic Standards Multi-Element Standards
Certified reference material (CRM) for ion chromatography (IC)
50 ppm Li+; 200 ppm Na+, K+, Mg²⁺; 400 ppm NH₄⁺, 1000 ppm Ca²⁺
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility