VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Supplier: Ricca Chemical
RV010685-50N RV010685-100N
RCRV010685-50NEA 684.14 CAD
RCRV010685-50N RCRV010685100N
VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Standards Anionic Standards Multi-Element Anioinic Standards
Certified reference material (CRM) for ion chromatography (IC)
150 ppm SO₄²-, PO₄³; 100 ppm NO₃-, Br-, NO₂-; 30 ppm Cl-, 20 ppm F-
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility