VeriSpec™ Manganese Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010602-100N
RCRV010602100NEA 224.04 CAD
RCRV010602100N
VeriSpec™ Manganese Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Manganese Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility