VeriSpec® Tungsten Standard for ICP/MS 100 ppm NH₄OH, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010565-100N
RCRV010565100NEA 251.22 CAD
RCRV010565100N
VeriSpec® Tungsten Standard for ICP/MS 100 ppm NH₄OH, RICCA Chemical Company
Standards Cationic Standards Tungsten Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility