VeriSpec™ Gadolinium Standard for ICP/MS 100 ppm in 2% HNO₃
Supplier: RICCA Chemical Company
VeriSpec™
RV010522-100N
RCRV010522100NEA 251.22 CAD
RCRV010522100N
VeriSpec™ Gadolinium Standard for ICP/MS 100 ppm in 2% HNO₃
Standards Cationic Standards Gadolinium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility