VeriSpec® Molybdenum Standard for ICP 10 g/L in 4% NH₃, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010466-250N RV010466-50N RV010466-100N RV010466-500N
RCRV010466500NEA 710.64 CAD
RCRV010466500N RCRV010466100N RCRV010466250N RCRV010466-50N
VeriSpec® Molybdenum Standard for ICP 10 g/L in 4% NH₃, Ricca Chemical Company
Standards Cationic Standards Molybdenum Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility