VeriSpec™ Potassium Standard for ICP 10 g/L in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010460-100N
RV010460-250N
RV010460-500N
RCRV010460-50NEA
141.52
CAD
RCRV010460-50N
RCRV010460100N
RCRV010460250N
RCRV010460500N
VeriSpec™ Potassium Standard for ICP 10 g/L in 2% HNO₃, RICCA Chemical Company
Standards
Cationic Standards
Potassium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 10 g/L= 10,000 µg/mL