VeriSpec® Barium Standard for ICP 10 g/L in 2% HNO₃
Supplier: Ricca Chemical
RV010438-100N RV010438-500N
RCRV010438-50NEA 113.21 CAD
RCRV010438-50N RCRV010438100N RCRV010438250N RCRV010438500N
VeriSpec® Barium Standard for ICP 10 g/L in 2% HNO₃
Standards Cationic Standards Barium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Order Now
SPECIFICATIONS
Environmentally Preferable