VeriSpec® Tungsten Standard for ICP 1000 ppm in 4% NH₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010360-100N
RV010360-250N
RV010360-500N
RCRV010360-50NEA
122.69
CAD
RCRV010360-50N
RCRV010360100N
RCRV010360250N
RCRV010360500N
VeriSpec® Tungsten Standard for ICP 1000 ppm in 4% NH₃, RICCA Chemical Company
Standards
Cationic Standards
Tungsten Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility