VeriSpec® Tellurium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010356-100N
RV010356-250N
RV010356-500N
RCRV010356-50NEA
565.96
CAD
RCRV010356-50N
RCRV010356100N
RCRV010356250N
RCRV010356500N
VeriSpec® Tellurium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards
Cationic Standards
Tellurium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility