VeriSpec® Rhenium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010350-100N RV010350-250N RV010350-500N
RCRV010350-50NEA 538.72 CAD
RCRV010350-50N RCRV010350100N RCRV010350250N RCRV010350500N
VeriSpec® Rhenium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Rhenium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility