VeriSpec® Phosphorus Standard for ICP 1000 ppm in H₂SO₄, RICCA Chemical Company
Supplier: Ricca Chemical
RV010346-100N RV010346-250N RV010346-500N
RCRV010346-50NEA 186.3 CAD
RCRV010346-50N RCRV010346100N RCRV010346250N RCRV010346500N
VeriSpec® Phosphorus Standard for ICP 1000 ppm in H₂SO₄, RICCA Chemical Company
Standards Cationic Standards Phosphorus Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility