VeriSpec® Chromium Standard for ICP 1000 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010336-100N RV010336-250N RV010336-500N
RCRV010336-50NEA 126.18 CAD
RCRV010336-50N RCRV010336100N RCRV010336250N RCRV010336500N
VeriSpec® Chromium Standard for ICP 1000 ppm in 2% HNO₃
Standards Cationic Standards Chromium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility