VeriSpec® Calcium Standard for ICP 1000 ppm in 2% HNO₃
Supplier: RICCA Chemical Company
VeriSpec™
RV010335-50N RV010335-100N RV010335-250N RV010335-500N
RCRV010335250NEA 335.17 CAD
RCRV010335250N RCRV010335100N RCRV010335500N RCRV010335-50N
VeriSpec® Calcium Standard for ICP 1000 ppm in 2% HNO₃
Standards Cationic Standards Calcium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility