VeriSpec® Beryllium Standard for ICP 1000 ppm in 2% HNO₃/HF, Ricca Chemical Company
Supplier: Ricca Chemical
RV010334-100N RV010334-250N RV010334-500N
RCRV010334-50NEA 249.28 CAD
RCRV010334-50N RCRV010334100N RCRV010334250N RCRV010334500N
VeriSpec® Beryllium Standard for ICP 1000 ppm in 2% HNO₃/HF, Ricca Chemical Company
Standards Cationic Standards Beryllium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Order Now
SPECIFICATIONS
Environmentally Preferable