VeriSpec® Barium Standard for ICP 1000 ppm in 2% HNO₃
Supplier: RICCA Chemical Company
VeriSpec™
RV010333-50N RV010333-500N RV010333-250N RV010333-100N
RCRV010333500NEA 485.22 CAD
RCRV010333500N RCRV010333100N RCRV010333250N RCRV010333-50N
VeriSpec® Barium Standard for ICP 1000 ppm in 2% HNO₃
Standards Cationic Standards Barium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility