VeriSpec™ Zinc Standard for ICP 1000 ppm in 2% HCl, RICCA Chemical Company
Supplier: Ricca Chemical
RV010330-100N RV010330-250N RV010330-500N
RCRV010330-50NEA 115.58 CAD
RCRV010330-50N RCRV010330100N RCRV010330250N RCRV010330500N
VeriSpec™ Zinc Standard for ICP 1000 ppm in 2% HCl, RICCA Chemical Company
Standards Cationic Standards Zinc Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility