VeriSpec™ Ytterbium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010329-100N RV010329-250N RV010329-500N
RCRV010329-50NEA 122.69 CAD
RCRV010329-50N RCRV010329100N RCRV010329250N RCRV010329500N
VeriSpec™ Ytterbium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Ytterbium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility