VeriSpec® Vanadium Standard for ICP 1000 ppm in 2% H₂SO₄, RICCA Chemical Company
Supplier: Ricca Chemical
RV010326-100N RV010326-250N RV010326-500N
RCRV010326-50NEA 115.58 CAD
RCRV010326-50N RCRV010326100N RCRV010326250N RCRV010326500N
VeriSpec® Vanadium Standard for ICP 1000 ppm in 2% H₂SO₄, RICCA Chemical Company
Standards Cationic Standards Vanadium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility