VeriSpec® Uranium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010325-100N RV010325-250N RV010325-500N
RCRV010325-50NEA 141.52 CAD
RCRV010325-50N RCRV010325100N RCRV010325250N RCRV010325500N
VeriSpec® Uranium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Uranium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Order Now
SPECIFICATIONS
Environmentally Preferable