VeriSpec® Thallium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010323-500N RV010323-250N RV010323-100N RV010323-50N
RCRV010323250NEA 334.85 CAD
RCRV010323250N RCRV010323100N RCRV010323-50N RCRV010323500N
VeriSpec® Thallium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Thallium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility