VeriSpec® Silicon Standard for ICP 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010313-100N RV010313-250N RV010313-500N
RCRV010313-50NEA 115.58 CAD
RCRV010313-50N RCRV010313100N RCRV010313250N RCRV010313500N
VeriSpec® Silicon Standard for ICP 1000 ppm in H₂O, RICCA Chemical Company
Standards Cationic Standards Silicium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility