VeriSpec® Phosphorus Standard for ICP 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010300-100N
RV010300-250N
RV010300-500N
RCRV010300-50NEA
115.58
CAD
RCRV010300-50N
RCRV010300100N
RCRV010300250N
RCRV010300500N
VeriSpec® Phosphorus Standard for ICP 1000 ppm in H₂O, RICCA Chemical Company
Standards
Cationic Standards
Phosphorus Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility