VeriSpec™ Nickel Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010298-100N RV010298-250N RV010298-500N
RCRV010298-50NEA 94.8 CAD
RCRV010298-50N RCRV010298100N RCRV010298250N RCRV010298500N
VeriSpec™ Nickel Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Nickel Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Order Now
SPECIFICATIONS
Environmentally Preferable