VeriSpec™ Manganese Standard for ICP 1000 ppm in 2% HCl, RICCA Chemical Company
Supplier: Ricca Chemical
RV010293-100N
RV010293-250N
RV010293-500N
RCRV010293100NEA
334.96
CAD
RCRV010293100N
RCRV010293250N
RCRV010293500N
VeriSpec™ Manganese Standard for ICP 1000 ppm in 2% HCl, RICCA Chemical Company
Standards
Cationic Standards
Manganese Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility