VeriSpec™ Dysprosium Standard for ICP 1000 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010276-100N
RV010276-250N
RV010276-500N
RCRV010276-50NEA
122.69
CAD
RCRV010276-50N
RCRV010276100N
RCRV010276250N
RCRV010276500N
VeriSpec™ Dysprosium Standard for ICP 1000 ppm in 2% HNO₃
Standards
Cationic Standards
Dysprosium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility