VeriSpec® Copper Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010275-100N RV010275-250N RV010275-500N
RCRV010275-50NEA 100.93 CAD
RCRV010275-50N RCRV010275100N RCRV010275250N RCRV010275500N
VeriSpec® Copper Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Copper Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility