VeriSpec® Cobalt Standard for ICP 1000 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010272-100N
RV010272-250N
RV010272-500N
RCRV010272100NEA
222.78
CAD
RCRV010272100N
RCRV010272250N
RCRV010272500N
VeriSpec® Cobalt Standard for ICP 1000 ppm in 2% HNO₃
Standards
Cationic Standards
Cobalt Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility