VeriSpec® Cadmium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010270-100N
RV010270-250N
RV010270-500N
RCRV010270-50NEA
115.58
CAD
RCRV010270-50N
RCRV010270100N
RCRV010270250N
RCRV010270500N
VeriSpec® Cadmium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards
Cationic Standards
Cadmium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility