VeriSpec® Barium Standard for ICP 1000 ppm in 2% HCl
Supplier: Ricca Chemical
RV010266-100N RV010266-250N RV010266-500N
RCRV010266-50NEA 115.58 CAD
RCRV010266-50N RCRV010266100N RCRV010266250N RCRV010266500N
VeriSpec® Barium Standard for ICP 1000 ppm in 2% HCl
Standards Cationic Standards Barium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility