VeriSpec® Strontium Ionization Buffer for AAS 2% Sr in 2% HCl, RICCA Chemical Company
Supplier: Ricca Chemical
RV010257-100N RV010257-500N
RCRV010257100NEA 881.64 CAD
RCRV010257100N RCRV010257500N
VeriSpec® Strontium Ionization Buffer for AAS 2% Sr in 2% HCl, RICCA Chemical Company
Standards Cationic Standards Strontium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility