VeriSpec™ Lithium Ionization Buffer for AAS 2% Li in 1% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010142-100N
RV010142-500N
RCRV010142100NEA
662.02
CAD
RCRV010142100N
RCRV010142500N
VeriSpec™ Lithium Ionization Buffer for AAS 2% Li in 1% HNO₃, RICCA Chemical Company
Standards
Cationic Standards
Lithium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility