VeriSpec™ Potassium (K) Standard for AAS 1000 ppm in 12% Ethanol, RICCA Chemical Company
Supplier: Ricca Chemical
RV010119-100N
RV010119-500N
RCRV010119100NEA
221.2
CAD
RCRV010119100N
RCRV010119500N
VeriSpec™ Potassium (K) Standard for AAS 1000 ppm in 12% Ethanol, RICCA Chemical Company
Standards
Cationic Standards
Potassium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility