VeriSpec® Vanadium (V) Standard for AAS 1000 ppm in 2% H₂SO₄, RICCA Chemical Company
Supplier: Ricca Chemical
RV010087-100N
RV010087-500N
RCRV010087100NEA
110.6
CAD
RCRV010087100N
RCRV010087500N
VeriSpec® Vanadium (V) Standard for AAS 1000 ppm in 2% H₂SO₄, RICCA Chemical Company
Standards
Cationic Standards
Vanadium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility