VeriSpec™ Lanthanum (La) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010076-100N RV010076-500N
RCRV010076100NEA 137.46 CAD
RCRV010076100N RCRV010076500N
VeriSpec™ Lanthanum (La) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Lanthanum Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility