VeriSpec™ Potassium (K) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010075-100N RV010075-500N
RCRV010075100NEA 110.6 CAD
RCRV010075100N RCRV010075500N
VeriSpec™ Potassium (K) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Potassium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility