VeriSpec® Copper (Cu) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010073-100N
RV010073-500N
RCRV010073-1NEA
228.06
CAD
RCRV010073-1N
RCRV010073100N
RCRV010073500N
VeriSpec® Copper (Cu) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards
Cationic Standards
Copper Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility