VeriSpec® Calcium (Ca) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010068-100N RV010068-500N
RCRV010068-1NEA 228.06 CAD
RCRV010068-1N RCRV010068100N RCRV010068500N
VeriSpec® Calcium (Ca) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Calcium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility