VeriSpec™ Yttrium (Y) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010060-100N RV010060-500N
RCRV010060100NEA 126.4 CAD
RCRV010060100N RCRV010060500N
VeriSpec™ Yttrium (Y) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Yttrium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
SPECIFICATIONS
Environmentally Preferable