VeriSpec® Vanadium (V) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010058-100N
RV010058-500N
RCRV010058100NEA
118.5
CAD
RCRV010058100N
RCRV010058500N
VeriSpec® Vanadium (V) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards
Cationic Standards
Vanadium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility