VeriSpec® Samarium (Sm) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010051-100N
RV010051-500N
RCRV010051100NEA
158
CAD
RCRV010051100N
RCRV010051500N
VeriSpec® Samarium (Sm) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards
Cationic Standards
Samarium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility