VeriSpec® Silicon (Si) Standard for AAS 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010050-100N RV010050-500N
RCRV010050-1NEA 364.81 CAD
RCRV010050-1N RCRV010050100N RCRV010050500N
VeriSpec® Silicon (Si) Standard for AAS 1000 ppm in H₂O, RICCA Chemical Company
Standards Cationic Standards Silicium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility