VeriSpec® Selenium (Se) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010049-100N
RV010049-500N
RCRV010049100NEA
120.08
CAD
RCRV010049100N
RCRV010049500N
VeriSpec® Selenium (Se) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards
Cationic Standards
Selenium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility