VeriSpec™ Magnesium (Mg) Standard for AAS 1000 ppm in 2% HCl, RICCA Chemical Company
Supplier: Ricca Chemical
RV010031-100N RV010031-500N
RCRV010031-1NEA 228.06 CAD
RCRV010031-1N RCRV010031100N RCRV010031500N
VeriSpec™ Magnesium (Mg) Standard for AAS 1000 ppm in 2% HCl, RICCA Chemical Company
Standards Cationic Standards Magnesium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Order Now
SPECIFICATIONS
Environmentally Preferable