VeriSpec™ Lithium (Li) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010030-100N RV010030-500N
RCRV010030100NEA 120.08 CAD
RCRV010030100N RCRV010030500N
VeriSpec™ Lithium (Li) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Lithium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility