VeriSpec™ Potassium (K) Standard for AAS 1000 ppm in 2% HCl, RICCA Chemical Company
Supplier: Ricca Chemical
RV010028-100N
RV010028-500N
RCRV010028100NEA
120.08
CAD
RCRV010028100N
RCRV010028500N
VeriSpec™ Potassium (K) Standard for AAS 1000 ppm in 2% HCl, RICCA Chemical Company
Standards
Cationic Standards
Potassium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility