VeriSpec® Indium (In) Standard for AAS 1000 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010026-100N RV010026-500N
RCRV010026100NEA 142.2 CAD
RCRV010026100N RCRV010026500N
VeriSpec® Indium (In) Standard for AAS 1000 ppm in 2% HNO₃
Standards Cationic Standards Indium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility